Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gur-3
Entrez Gene ID
181245
Gene Name
Gustatory receptor family protein 3
Gene Aliases
CELE_ZC504.5
Chromosome Location
Chr.X: 10402522 - 10406303 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077342.6 NP_509743.2 9 - 10 1150 103

View Full Product Details