Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- egl-36
- Entrez Gene ID
-
181269
- Gene Name
- Potassium voltage-gated channel protein egl-36
- Gene Aliases
-
CELE_R07A4.1
- Chromosome Location
-
Chr.X: 10734326 - 10744355 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_077394.4 |
NP_509795.1 |
1 - 2 |
85 |
66 |
View Full Product Details