Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
egl-36
Entrez Gene ID
181269
Gene Name
Potassium voltage-gated channel protein egl-36
Gene Aliases
CELE_R07A4.1
Chromosome Location
Chr.X: 10734326 - 10744355 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077394.4 NP_509795.1 6 - 7 723 63

View Full Product Details