Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ttr-14
Entrez Gene ID
181272
Gene Name
TransThyretin-Related family domain
Gene Aliases
CELE_T05A10.3
Chromosome Location
Chr.X: 10787503 - 10789700 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077400.6 NP_509801.1 7 - 8 927 84

View Full Product Details