Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nas-10
Entrez Gene ID
181287
Gene Name
Metalloendopeptidase
Gene Aliases
CELE_K09C8.3
Chromosome Location
Chr.X: 10960914 - 10963507 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270197.3 NP_001257126.1 9 - 10 1583 80

View Full Product Details