Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clc-1
Entrez Gene ID
181293
Gene Name
CLaudin-like in Caenorhabditis
Gene Aliases
CELE_C09F12.1
Chromosome Location
Chr.X: 11047446 - 11049275 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077446.6 NP_509847.1 2 - 3 259 142

View Full Product Details