Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
npr-9
Entrez Gene ID
191320
Gene Name
Galanin-like G-protein coupled receptor npr-9
Gene Aliases
CELE_ZK455.3
Chromosome Location
Chr.X: 11329724 - 11332169 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001375132.1 NP_001361821.1 5 - 6 731 68

View Full Product Details