Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
prx-12
Entrez Gene ID
181331
Gene Name
Putative peroxisome assembly protein 12
Gene Aliases
CELE_F08B12.2
Chromosome Location
Chr.X: 11393293 - 11394619 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322631.2 NP_001309444.1 5 - 6 987 87

View Full Product Details