Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
set-6
Entrez Gene ID
181371
Gene Name
SET domain-containing protein
Gene Aliases
CELE_C49F5.2
Chromosome Location
Chr.X: 11970030 - 11974679 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077602.5 NP_510003.2 7 - 8 1096 97

View Full Product Details