Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
xtr-2
Entrez Gene ID
192058
Gene Name
protein-coding
Gene Aliases
CELE_F17A2.1
Chromosome Location
Chr.X: 12315568 - 12318184 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322636.1 NP_001309688.1 1 - 2 53 99
NM_077658.4 NP_510059.2 1 - 2 390 99

View Full Product Details