Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ceh-40
Entrez Gene ID
191624
Gene Name
Homeobox protein ceh-40
Gene Aliases
CELE_F17A2.5
Chromosome Location
Chr.X: 12318126 - 12320777 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077659.4 NP_510060.1 5 - 6 1020 81

View Full Product Details