Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
atf-6
Entrez Gene ID
181405
Gene Name
BZIP domain-containing protein
Gene Aliases
CELE_F45E6.2
Chromosome Location
Chr.X: 12480948 - 12484912 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077693.6 NP_510094.1 5 - 6 910 77

View Full Product Details