Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-10
Entrez Gene ID
187547
Gene Name
protein-coding
Gene Aliases
CELE_R03G8.5
Chromosome Location
Chr.X: 13082626 - 13084235 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322638.1 NP_001309672.1 1 - 2 199 111
NM_001322639.1 NP_001309673.1 1 - 2 199 111

View Full Product Details