Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-32
Entrez Gene ID
187178
Gene Name
Nuclear Hormone Receptor family
Gene Aliases
CELE_K08H2.8
Chromosome Location
Chr.X: 13251964 - 13256289 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077800.7 NP_510201.2 3 - 4 367 87

View Full Product Details