Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-13B1
Entrez Gene ID
181462
Gene Name
Putative cytochrome P450 cyp-13B1
Gene Aliases
CELE_F02C12.5
Chromosome Location
Chr.X: 13406674 - 13409704 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077832.5 NP_510233.1 7 - 8 1245 136

View Full Product Details