Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
meg-1
Entrez Gene ID
181502
Gene Name
Uncharacterized protein
Gene Aliases
CELE_K02B9.1
Chromosome Location
Chr.X: 13927036 - 13929634 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077918.8 NP_510319.1 4 - 5 1762 121

View Full Product Details