Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ceh-36
Entrez Gene ID
181529
Gene Name
Homeobox protein ceh-36
Gene Aliases
CELE_C37E2.4
Chromosome Location
Chr.X: 14183221 - 14186566 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077964.5 NP_510365.1 2 - 3 313 87

View Full Product Details