Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
prx-1
Entrez Gene ID
181538
Gene Name
protein-coding
Gene Aliases
CELE_C11H1.4
Chromosome Location
Chr.X: 14313721 - 14326726 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077985.6 NP_510386.2 4 - 5 696 96
GenBank mRNA AB054992.1 - 4 - 5 696 96

View Full Product Details