Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- B0222.11
- Entrez Gene ID
-
3896831
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_B0222.11
- Chromosome Location
-
Chr.V: 9159678 - 9168189 on Build WBcel235
- Gene Symbol
- col-10
- Entrez Gene ID
-
179298
- Gene Name
- Cuticle collagen 10
- Gene Aliases
-
CELE_B0222.8
- Chromosome Location
-
Chr.V: 9159667 - 9165437 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383367.1 |
|
1 - 2 |
258 |
103 |
View Full Product Details