Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mlc-3
Entrez Gene ID
175768
Gene Name
protein-coding
Gene Aliases
CELE_F09F7.2
Chromosome Location
Chr.III: 5565025 - 5567382 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001372589.2 NP_001359961.1 2 - 2 327 74

View Full Product Details