Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- pnc-2
- Entrez Gene ID
-
3565064
- Gene Name
- Isochorismatase domain-containing protein
- Gene Aliases
-
CELE_Y57G11C.47
- Chromosome Location
-
Chr.IV: 14904978 - 14909689 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383250.1 |
|
4 - 5 |
719 |
102 |
View Full Product Details