Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pnc-2
Entrez Gene ID
3565064
Gene Name
Isochorismatase domain-containing protein
Gene Aliases
CELE_Y57G11C.47
Chromosome Location
Chr.IV: 14904978 - 14909689 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383250.1 7 - 8 1078 64

View Full Product Details