Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dlc-5
Entrez Gene ID
3565343
Gene Name
Dynein Light Chain
Gene Aliases
CELE_Y73B6BL.43
Chromosome Location
Chr.IV: 6438204 - 6440032 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028400.3 NP_001023571.1 1 - 2 141 85

View Full Product Details