Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dlc-5
Entrez Gene ID
3565343
Gene Name
Dynein Light Chain
Gene Aliases
CELE_Y73B6BL.43
Chromosome Location
Chr.IV: 6438204 - 6440032 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028400.3 NP_001023571.1 3 - 4 389 127

View Full Product Details