Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srz-44
Entrez Gene ID
3565821
Gene Name
Serpentine Receptor, class Z
Gene Aliases
CELE_C55A1.12
Chromosome Location
Chr.V: 15637416 - 15638953 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028567.1 NP_001023738.1 3 - 4 550 115

View Full Product Details