Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srsx-10
Entrez Gene ID
3565803
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_F26G5.12
Chromosome Location
Chr.V: 4973235 - 4974777 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028692.3 NP_001023863.1 1 - 2 134 150

View Full Product Details