Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-82
Entrez Gene ID
3565043
Gene Name
7TM_GPCR_Srx domain-containing protein
Gene Aliases
CELE_F43A11.6
Chromosome Location
Chr.V: 11589733 - 11592931 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028745.1 NP_001023916.1 3 - 4 406 186

View Full Product Details