Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxc-48
Entrez Gene ID
182425
Gene Name
F-box C protein
Gene Aliases
CELE_C08G5.2
Chromosome Location
Chr.II: 739553 - 740908 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061399.6 NP_493800.3 1 - 2 130 75

View Full Product Details