Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hhat-1
Entrez Gene ID
259478
Gene Name
Homolog of Hedgehog AcylTransferase
Gene Aliases
CELE_ZC101.3
Chromosome Location
Chr.II: 14678457 - 14681148 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027318.6 NP_001022489.1 7 - 8 1520 88

View Full Product Details