Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srbc-71
Entrez Gene ID
3564793
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_F30B5.8
Chromosome Location
Chr.IV: 4222177 - 4225970 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027991.2 NP_001023162.1 1 - 2 200 149

View Full Product Details