Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
glb-21
Entrez Gene ID
3565100
Gene Name
GLOBIN domain-containing protein
Gene Aliases
CELE_R102.9
Chromosome Location
Chr.IV: 10709548 - 10711794 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028168.3 NP_001023339.1 1 - 2 100 71

View Full Product Details