Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srv-28
Entrez Gene ID
3565289
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_T13A10.14
Chromosome Location
Chr.IV: 6261058 - 6263344 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028212.1 NP_001023383.1 5 - 6 881 155

View Full Product Details