Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-80
Entrez Gene ID
3565458
Gene Name
protein-coding
Gene Aliases
CELE_Y54G2A.39
Chromosome Location
Chr.IV: 2823746 - 2828069 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028333.4 NP_001023504.1 1 - 2 150 89
NR_132458.1 1 - 2 90 89

View Full Product Details