Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srz-13
Entrez Gene ID
3565887
Gene Name
Serpentine Receptor, class Z
Gene Aliases
CELE_K05D4.8
Chromosome Location
Chr.V: 16182410 - 16183540 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028859.1 NP_001024030.1 2 - 3 749 143

View Full Product Details