Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MT4
Entrez Gene ID
403769
Gene Name
metallothionein 4
Gene Aliases
-
Chromosome Location
Chr.2: 59640784 - 59643370 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003150.1 NP_001003150.1 1 - 2 68 91
GenBank mRNA AB028041.1 - 1 - 2 68 91

View Full Product Details