Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL4
Entrez Gene ID
403785
Gene Name
interleukin 4
Gene Aliases
IL-4
Chromosome Location
Chr.11: 20972693 - 20981539 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003159.1 NP_001003159.1 1 - 1 120 110
GenBank mRNA AF054833.1 - 1 - 1 57 110
AF072696.1 - 1 - 1 57 110
AF083270.1 - 1 - 1 99 110
AF104245.1 - 1 - 1 57 110
AF187322.1 - 1 - 1 123 110
AF239917.1 - 1 - 1 120 110
AF328928.1 - 1 - 1 43 110
EF095771.1 - 1 - 1 67 110

View Full Product Details