Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SOX9
Entrez Gene ID
403464
Gene Name
SRY-box transcription factor 9
Gene Aliases
-
Chromosome Location
Chr.9: 8275049 - 8278172 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002978.1 NP_001002978.1 - 683 103
GenBank mRNA AY237827.1 - - 683 103

View Full Product Details