Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP1CA
Entrez Gene ID
403609
Gene Name
protein phosphatase 1 catalytic subunit alpha
Gene Aliases
-
Chromosome Location
Chr.18: 50062352 - 50065166 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003064.2 NP_001003064.2 2 - 3 287 81
GenBank mRNA AY062037.1 - 2 - 3 193 81
DN365927.1 - 2 - 3 205 81
DN392813.1 - 2 - 3 296 81
DN871864.1 - 2 - 3 204 81

View Full Product Details