Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZP3
Entrez Gene ID
403894
Gene Name
zona pellucida glycoprotein 3
Gene Aliases
ZPC, Zp-3
Chromosome Location
Chr.6: 7586212 - 7594610 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003224.1 NP_001003224.1 - 334 74
GenBank mRNA D45070.1 - - 334 74
U05780.1 - - 307 74

View Full Product Details