Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPCS3
Entrez Gene ID
404005
Gene Name
signal peptidase complex subunit 3
Gene Aliases
SPC22/23
Chromosome Location
Chr.16: 53258714 - 53266191 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003314.1 NP_001003314.1 3 - 4 442 114
GenBank mRNA J04067.1 - 3 - 4 442 114

View Full Product Details