Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LGALS3
Entrez Gene ID
404021
Gene Name
galectin 3
Gene Aliases
CBP 35, Gal-3, Galectin-3
Chromosome Location
Chr.8: 30977438 - 30994711 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197043.1 NP_001183972.1 2 - 3 458 78
XM_005623184.3 XP_005623241.1 3 - 4 594 78
GenBank mRNA L23429.1 - 1 - 2 447 78

View Full Product Details