Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP2B6
Entrez Gene ID
474177
Gene Name
cytochrome P450 2B11
Gene Aliases
CYP2B11, CYPIIB11
Chromosome Location
Chr.1: 112817561 - 112833294 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006652.1 NP_001006653.1 3 - 4 495 66
GenBank mRNA HG965162.1 - 3 - 4 487 66
HG965163.1 - 3 - 4 487 66
HG974426.1 - 3 - 4 487 66
M92447.1 - 3 - 4 495 66

View Full Product Details