Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTD
Entrez Gene ID
477059
Gene Name
biotinidase
Gene Aliases
-
Chromosome Location
Chr.23: 27037640 - 27069544 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005634391.3 XP_005634448.1 4 - 5 698 81
XM_005634393.3 XP_005634450.1 5 - 6 805 81
XM_022408610.1 XP_022264318.1 3 - 4 505 81

View Full Product Details