Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRSN1
Entrez Gene ID
478734
Gene Name
neurensin 1
Gene Aliases
-
Chromosome Location
Chr.35: 22216835 - 22237398 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_535901.6 XP_535901.3 3 - 4 443 77

View Full Product Details