Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHURC1
Entrez Gene ID
609942
Gene Name
churchill domain containing 1
Gene Aliases
-
Chromosome Location
Chr.8: 39301090 - 39318352 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_847755.5 XP_852848.2 1 - 2 128 56

View Full Product Details