Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAMTOR2
Entrez Gene ID
480125
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 2
Gene Aliases
ROBLD3
Chromosome Location
Chr.7: 41760632 - 41764926 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005622713.3 XP_005622770.1 1 - 2 209 87
XM_537247.6 XP_537247.1 1 - 2 200 87

View Full Product Details