Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- AIMP2
- Entrez Gene ID
-
479752
- Gene Name
- aminoacyl tRNA synthetase complex interacting multifunctional protein 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.6: 11438768 - 11446307 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_536880.6 |
XP_536880.2 |
- |
278 |
96 |
View Full Product Details