Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APOA1
Entrez Gene ID
479427
Gene Name
apolipoprotein A1
Gene Aliases
Apo-AI, ApoA-I
Chromosome Location
Chr.5: 16740071 - 16742081 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014113398.2 XP_013968873.1 - 156 61

View Full Product Details