Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCR9
Entrez Gene ID
484793
Gene Name
C-C motif chemokine receptor 9
Gene Aliases
-
Chromosome Location
Chr.20: 42607024 - 42616823 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284476.1 NP_001271405.1 2 - 3 89 80
GenBank mRNA AB622806.1 - 2 - 3 89 80

View Full Product Details