Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APEX1
Entrez Gene ID
482558
Gene Name
apurinic/apyrimidinic endodeoxyribonuclease 1
Gene Aliases
-
Chromosome Location
Chr.15: 17786808 - 17789422 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145119.1 NP_001138591.1 3 - 4 339 61

View Full Product Details